B-Tribromoborazine, 96%
B-Tribromoborazine, 96%
B-Tribromoborazine, 96%
Thermo Scientific Chemicals

B-Tribromoborazine, 96%

CAS: 13703-88-3 | B3Br3H3N3 | 317.187 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 043723.06
also known as 043723-06
Price (USD)
387.65
Online Exclusive
431.00
Save 43.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
387.65
Online Exclusive
431.00
Save 43.35 (10%)
Each
Chemical Identifiers
CAS13703-88-3
IUPAC Name2,4,6-tribromo-1,3,5,2,4,6-triazatriborinane
Molecular FormulaB3Br3H3N3
InChI KeyPCLMMJIWMGUYKF-UHFFFAOYSA-N
SMILESBrB1NB(Br)NB(Br)N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point125.0-129.0°C
Appearance (Color)White
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only