Thermo Scientific Chemicals

Niobium(V) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043729.06
5 g, Each
Thermo Scientific Chemicals

Niobium(V) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043729.06
5 g, Each
Quantity
Catalog number: 043729.06
also known as 043729-06
Price (USD)
Price: 113.00
Online price: 96.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10026-12-7
IUPAC Name
niobium(5+) pentachloride
Molecular Formula
Cl5Nb
InChI Key
YHBDIEWMOMLKOO-UHFFFAOYSA-I
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Nb+5]
Total Metal Impurities
0.001% max.

Description

Niobium(V) chloride is the precursor in the preparation of pure niobium, and ferroniobium. It is used as a Lewis acid catalyst in organic synthesis in activating alkenes in the carbonyl-ene reaction and the Diels-Alder reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text