Thermo Scientific Chemicals

Titanium(IV) oxide, catalyst support, Thermo Scientific Chemicals

Catalog number: 043828.A7
5 kg, Each
Thermo Scientific Chemicals

Titanium(IV) oxide, catalyst support, Thermo Scientific Chemicals

Catalog number: 043828.A7
5 kg, Each
Quantity
Catalog number: 043828.A7
also known as 043828-A7
Price (USD)
Price: 648.00
Online price: 560.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1317-70-0
IUPAC Name
dioxotitanium
Molecular Formula
O2Ti
InChI Key
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILES
O=[Ti]=O
Comment
Crush strength: ≥4lb
Comment
Bulk density: 50-80lb/ft³
Comment
Total pore volume: 0.20-0.40cc/g
Surface Area
30-50m²/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text