Aluminum oxide, gamma-phase, catalyst support, high surface area, bimodal
Aluminum oxide, gamma-phase, catalyst support, high surface area, bimodal
Aluminum oxide, gamma-phase, catalyst support, high surface area, bimodal
Aluminum oxide, gamma-phase, catalyst support, high surface area, bimodal
Aluminum oxide, gamma-phase, catalyst support, high surface area, bimodal
Thermo Scientific Chemicals

Aluminum oxide, gamma-phase, catalyst support, high surface area, bimodal

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 043832.30
also known as 043832-30
Price (EUR)
74,38
Online Exclusive
87,50
Save 13,12 (15%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (EUR)
74,38
Online Exclusive
87,50
Save 13,12 (15%)
Each
Add to cart
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
CommentTotal pore volume, (Hg): 0.80 to 1.20 cc/g
CommentPacking density: 325 to 475 kg/m³
Surface Area220 to 280 m²/g
FormPellets
Aluminum oxide finds applications in catalyst support, chemical sensors, as an absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs and heat-resistant fiber. Heavy quantity of Aluminum oxide is used in the production of aluminum and a component of clay. It is also used as catalyst in hydrodesulfurization and in Ziegler-Natta polymerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only