Thermo Scientific Chemicals

Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals

Catalog number: 043902.22
100 g, Each
Thermo Scientific Chemicals

Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals

Catalog number: 043902.22
100 g, Each
Quantity
Catalog number: 043902.22
also known as 043902-22
Price (USD)
Price: 142.00
Online price: 120.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13586-38-4
Melting Point
120-124
Appearance (Color)
Pink to Red
Assay (unspecified)
99% min
Form
Powder or Crystals

Description

Ammonium cobalt(II) sulfate hexahydrate is uses as spectroscopic standard, cobalt plating, catalysis and ceramics. It is also used in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text