N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS
Thermo Scientific Chemicals

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS

CAS: 25561-30-2 | C8H18F3NOSi2 | 257.403 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
5 x 1 g
100 g
Catalog number 043939.46
also known as 043939-46
Price (TWD)
-
Quantity:
5 x 1 g
Request bulk or custom format
Chemical Identifiers
CAS25561-30-2
IUPAC Nametrimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)ethanimidate
Molecular FormulaC8H18F3NOSi2
InChI KeyXCOBLONWWXQEBS-UHFFFAOYSA-N
SMILESC[Si](C)(C)OC(=N[Si](C)(C)C)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)≤10
Assay (GC)0.5-4.0% TMCS
FormLiquid
Assay (GC)≥93.0%
Appearance (Color)Clear
N,O-Bis(trimethylsilyl)trifluoroacetamide is used as a derivatization reagent for gas chromatography and mass-spectrometry analysis of phenolic acids in fruits. It is also used as a reactant in the preparation of pyrimidinone ribosides and analogs which shows anti-tumor properties. It plays as an important role as a reagent for the regioselective desulfation of polysaccharide sulfates. Furthermore, it is used to prepare trimethylsilyl ethers of carbohydrates and alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only