Tantalum gauze, 30 mesh woven from 0.076mm (0.003in) dia wire
Tantalum gauze, 30 mesh woven from 0.076mm (0.003in) dia wire
Tantalum gauze, 30 mesh woven from 0.076mm (0.003in) dia wire
Thermo Scientific Chemicals

Tantalum gauze, 30 mesh woven from 0.076mm (0.003in) dia wire

CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
125 x 125 mm
250 x 250 mm
Catalog number 043965.GN
also known as 043965-GN
Price (EUR)
-
Quantity:
125 x 125 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
FormGauze
Comment30 mesh woven from 0.076mm dia wire
Appearance (Color)Gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only