Thermo Scientific Chemicals

Magnesium oxalate dihydrate, 98.5+%, Thermo Scientific Chemicals

Catalog number: 043976.36
500 g, Each
Thermo Scientific Chemicals

Magnesium oxalate dihydrate, 98.5+%, Thermo Scientific Chemicals

Catalog number: 043976.36
500 g, Each
Quantity
Catalog number: 043976.36
also known as 043976-36
Price (USD)
Price: 183.00
Online price: 158.65
Your price:
Quantity
-

Chemical Identifiers

CAS
547-66-0
IUPAC Name
magnesium(2+) oxalate
Molecular Formula
C2MgO4
InChI Key
UHNWOJJPXCYKCG-UHFFFAOYSA-L
SMILES
[Mg++].[O-]C(=O)C([O-])=O
Assay (unspecified)
98.5% min

Description

Magnesium oxalate dihydrate is in the synthesis of nano sized magnesium oxide. It is also used as a catalyst, refractory materials, adsorbents, superconductors, and ferroelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text