Poly(methyl methacrylate)
Poly(methyl methacrylate)
Poly(methyl methacrylate)
Poly(methyl methacrylate)
Thermo Scientific Chemicals

Poly(methyl methacrylate)

CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 043982.18
also known as 043982-18
Price (EUR)
51,60
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
51,60
Each
Chemical Identifiers
CAS9011-14-7
Molecular Formula(C5H8O2)n
SMILESCOC(=O)C(C)(-*)C-*
MDL NumberMFCD00134349
SpecificationsSpecification SheetSpecification Sheet
Formcrystalline powder
Particle Size-160 mesh /+100 mesh
Appearance (Color)White
Molecular Weight400,000 to 550,000
Poly(methyl methacrylate) is used in the lenses of exterior lights of automobile and automotive components such as badges, steering wheel, tachometer covers, warning triangles and fascia panels. It is in the form of sheets used in shatter resistant panels for building windows, skylights, bullet proof security barriers, bathtubs, LCD screens, furniture and many other applications. It is used as the matrix in laser dye-doped organic solid-state gain media for tunable solid state dye lasers. Further, it is used as a shield to stop beta radiation emitted from radioisotopes, as dosimeter devices during the gamma irradiation process and as a dispersant for ceramic powders to stabilize colloidal suspensions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only