Ethanol, denatured, Spectrophotometric Grade, 90%, packaged under Argon in resealable ChemSeal™ bottles
Ethanol, denatured, Spectrophotometric Grade, 90%, packaged under Argon in resealable ChemSeal™ bottles
Ethanol, denatured, Spectrophotometric Grade, 90%, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

Ethanol, denatured, Spectrophotometric Grade, 90%, packaged under Argon in resealable ChemSeal™ bottles

CAS: 64-17-5 | C2H6O | 46.069 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044014.K2
also known as 044014-K2
Price (TWD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS64-17-5
IUPAC Nameethanol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILESCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water0.1%
Residue on Evaporation5ppm
Ethanol, anhydrous, denatured is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For Manufacturing and Laboratory Use. Not for beverage use