Thermo Scientific Chemicals

Aluminum oxide desiccant, regular, Thermo Scientific Chemicals

Catalog number: 044018.36
500 g, Each
Thermo Scientific Chemicals

Aluminum oxide desiccant, regular, Thermo Scientific Chemicals

Catalog number: 044018.36
500 g, Each
Quantity
Catalog number: 044018.36
also known as 044018-36
Price (USD)
Quantity
-

Chemical Identifiers

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
Comment
3.2mm Spheres

Description

Aluminum oxide is used in dessicators ceramics, coating material and electrical insulators. It is an ingredient in sunscreen, sometimes present in cosmetics such as blush, lipstick, and nail polish. Aluminum oxide is the catalyst in the Claus process in hydro desulfurization and some Ziegler-Natta polymerizations. It is widely used as an abrasive, in the automotive industries, cosmetic industries and fiber materials.It is a medium for chromatography in some sodium vapor lamps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Dry gases and liquids
  • Gases include: CO2, acetylene, air, Ar, ethylene, He, H2, N2, O2
  • Liquids include: benzene, hexane, oil, pentane
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text