Dionex™ IonPac™ CS12 IC Columns
Actual product may vary
Thermo Scientific™

Dionex™ IonPac™ CS12 IC Columns

Green features
High-capacity, cation-exchange column for fast isocratic separation of common inorganic cations in diverse matrices.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
044019250 mm2 mmAnalytical Column
04400250 mm4 mmGuard Column
04402050 mm2 mmGuard Column
044001250 mm4 mmAnalytical Column
Catalog number 044019
Price (USD)
3,130.00
Each 1
Length (Metric):
250 mm
Diameter (Metric):
2 mm
Column Type:
Analytical Column
Price (USD)
3,130.00
Each 1
Obtain robust, reliable results and fast isocratic separation of common inorganic cations and alkanolamines using the Thermo Scientific™ Dionex™ IonPac™ CS12 Cation-Exchange Column. This is a high-capacity, cation-exchange column designed for the fast, isocratic separation of lithium, sodium, ammonium, potassium, magnesium, and calcium using methanesulfonic or sulfuric acid eluents in diverse sample matrices. However, the newer Thermo Scientific™ Dionex™ IonPac™ CS12A column provides improved performance and is the recommended column for inorganic cation analysis.
Fast Isocratic Separation of Common Inorganic Cations

The Dionex IonPac CS12 Cation-Exchange Column provides fast, isocratic separation of cations in diverse sample matrices, including drinking water, power plant waters, soil extracts, acid digests, chemical additives, chemical process solutions, scrubber solutions, and plating baths. However, the Dionex IonPac CS12A column improves performance for fast analysis of inorganic cations and ammonium in diverse matrices and is the recommend column for these applications. The columns are compatible with organic solvents, excluding alcohols, to enhance analyte solubility, modify column selectivity, or allow effective column cleanup

Multiple Formats Available for Application Flexibility
  • 4x250mm, 2x250mm analytical columns
  • 4x50mm, 2x50mm guard columns
  • Longer 250mm columns facilitate higher resolution separations of more complex sample matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column Capacity0.7 meq
Column ChemistryAcidic
Crosslinking55% DVB
Flow Rate0.25 mL/min.
For Use With (Application)Cation Exchange
For Use With (Equipment)Ion Chromatography Systems
Green FeaturesFSC
Max. Pressure4000 psi (275 bar)
Packing MaterialEthylvinylbenzene/Divinylbenzene
Particle Size8.5 μm
Pore Size60 &‌Aring;
QuantityEach 1
Solvent CompatibilityAcidic Eluents, 0 to 100% HPLC Solvent Compatible, Alcohols Should be Avoided
Solvents0 to 100 % HPLC Solvent Compatible. Alcohols Should be Avoided.
Stationary PhaseMacroporous Resin
Column TypeAnalytical Column
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)250 mm
Unit SizeEach 1