Formamide, Spectrophotometric Grade, 99+%
Formamide, Spectrophotometric Grade, 99+%
Formamide, Spectrophotometric Grade, 99+%
Formamide, Spectrophotometric Grade, 99+%
Thermo Scientific Chemicals

Formamide, Spectrophotometric Grade, 99+%

CAS: 75-12-7 | CH3NO | 45.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
4 x 500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044066.AP
also known as 044066-AP
Price (EUR)
195,00
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
195,00
Each
Add to cart
Chemical Identifiers
CAS75-12-7
IUPAC Nameformamide
Molecular FormulaCH3NO
InChI KeyZHNUHDYFZUAESO-UHFFFAOYSA-N
SMILESNC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Water Content (Karl Fischer Titration)≤ 0.03 %
Specific Absorbance (E 1%/1cm)300nm ≤ 0.05
Specific Absorbance (E 1%/1cm)400nm ≤ 0.01
Assay (GC)≥99.0 %
View more
Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only