Thermo Scientific™

Cobalt powder, -400 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

250 g, Each
Thermo Scientific™

Cobalt powder, -400 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

250 g, Each
Quantity
Catalog number: 044085.30
also known as 044085-30
Price (USD)
Price: 412.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
7440-48-4
IUPAC Name
cobalt
Molecular Formula
Co
InChI Key
GUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES
[Co]
Form
Powder, -400 mesh
Total Metal Impurities
≤0.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only