3,4,9,10-Perylenetetracarboxylic diimide
3,4,9,10-Perylenetetracarboxylic diimide
3,4,9,10-Perylenetetracarboxylic diimide
Thermo Scientific Chemicals

3,4,9,10-Perylenetetracarboxylic diimide

CAS: 81-33-4 | C24H10N2O4 | 390.354 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 044098.14
also known as 044098-14
Price (EUR)
198,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
198,00
Each
Chemical Identifiers
CAS81-33-4
IUPAC Name7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Molecular FormulaC24H10N2O4
InChI KeyKJOLVZJFMDVPGB-UHFFFAOYSA-N
SMILESO=C1NC(=O)C2=CC=C3C4=CC=C5C(=O)NC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Purple red
FormPowder
Assay from Supplier's CofA≥94%
Used in chemical research. PTCDI is used extensively as an industrial pigment. It is used as tunable laser dye, light-harvesting material, transistor, molecular switched, solar cell, and optoelectronic device. Planar perylene- and naphthalene-based diimide linkers can be employed to tether the Watson- Crick and the Hoogsteen strands of a DNA triplex, thus providing conjugates capable of targeting singlestranded nucleic acids with the formation of hairpin triplexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only