Vanadium foil, 0.075mm (0.003in) thick, 99.8% (metals basis)
Vanadium foil, 0.075mm (0.003in) thick, 99.8% (metals basis)
Vanadium foil, 0.075mm (0.003in) thick, 99.8% (metals basis)
Thermo Scientific Chemicals

Vanadium foil, 0.075mm (0.003in) thick, 99.8% (metals basis)

CAS: 7440-62-2 | V | 50.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 100 mm
100 x 200 mm
Catalog number 044106.GK
also known as 044106-GK
Price (EUR)
661,00
Each
Quantity:
100 x 200 mm
Request bulk or custom format
Price (EUR)
661,00
Each
Chemical Identifiers
CAS7440-62-2
IUPAC Namevanadium
Molecular FormulaV
InChI KeyLEONUFNNVUYDNQ-UHFFFAOYSA-N
SMILES[V]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White silver
FormFoil, 0.075mm (0.003in) thick.
Assay from Supplier's CofA≥99.8% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only