Offer consistent results with very good lifetimes with Thermo Scientific™ Dionex™ ED50A Electrochemical Detector Working Electrodes.
Description | ED50 A Working Electrode, Ag, with gasket and polishing pad |
For Use With (Equipment) | ED50 Electrochemical Detector |
Material | Silver in a Polymeric Support |
Product Line | Dionex |
Type | Silver Electrode |
Unit Size | Each |
Description | ED50 A Working Electrode, Au, with gasket and polishing pad |
For Use With (Equipment) | ED50 Electrochemical Detector |
Material | Gold in a Polymeric Support |
Product Line | Dionex |
Type | Gold Electrode |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
044114 | Each | ED50 A Working Electrode, Ag, with gasket and polishing pad | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
044112 also known as 44112 | Each | ED50 A Working Electrode, Au, with gasket and polishing pad | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Four types of electrode materials are available