Dionex™ IonPac™ CS14 IC Columns
Actual product may vary
Dionex™ IonPac™ CS14 IC Columns
Dionex™ IonPac™ CS14 IC Columns
Thermo Scientific™

Dionex™ IonPac™ CS14 IC Columns

Green features
Moderately low capacity cation-exchange column for analysis of aliphatic amines, aromatic amines, and polyamines using MSA or sulfuric acid eluents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
04412250 mm2 mmGuard Column
044121250 mm2 mmAnalytical Column
044123250 mm4 mmAnalytical Column
04412450 mm4 mmGuard Column
Catalog number 044122
Price (MXN)
-
Length (Metric):
50 mm
Diameter (Metric):
2 mm
Column Type:
Guard Column

Obtain consistent reliable results for the analysis of aliphatic amines, aromatic amines, and polyamines using methanesulfonic acid (MSA) or sulfuric acid eluents with the Thermo Scientific™ Dionex™ IonPac™ CS14 Cation-Exchange Column. The column facilitates resolution of a large number of aliphatic amines and Group I and Group II cations in diverse sample matrices. However, the newer Thermo Scientific™ Dionex™ IonPac™ CS17, CS19 and CS19-4μm cation-exchange columns are recommended for these separations.

Sample matrices can include environmental waters; power plant waters treated with ammonium, morpholine or ethanolamine; chemical additives; chemical process solutions; scrubber solutions; plating baths; industrial solvents; and wastewater.

Determination of Amines in Diverse Matrices
  • Medium low-capacity carboxylate-functionalized hydrophilic cation-exchange column optimized for determination of a variety of amines
  • Dionex IonPac CS19 and CS17 columns are recommended replacement columns for most Dionex IonPac CS14 column applications
  • Dionex IonPac CS19 and CS19-4μm columns are recommended for polar amines, including alkanolamines and methylamines; and moderately hydrophobic and polyvalent amines, including biogenic amines and alkyldiamines
  • Compatible with organic solvents, (THF up to 20%), to enhance analyte solubility, modify column selectivity, or allow effective column cleanup
Multiple Formats Available for Application Flexibility
  • 4x250mm, 2x250mm analytical columns
  • 4x50mm, 2x50mm guard columns
  • Longer 250mm columns facilitate higher resolution separations of more complex sample matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Application)Analysis of Aliphatic Amines and Polyamines
For Use With (Equipment)Analysis of Aliphatic Amines and Polyamines
Green FeaturesFSC
Max. Pressure4000 psi (275 bar)
Particle Size8 μm
QuantityEach 1
Solvent CompatibilityAcidic Eluents, 0 to 100% HPLC Solvents
Stationary PhaseIonPac CG14
Column TypeGuard Column
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)50 mm
Unit SizeEach 1