Thermo Scientific Chemicals

Ethanol, anhydrous, denatured, HPLC Grade, 90%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

100 mL, ChemSeal™ Bottle, Each
Thermo Scientific Chemicals

Ethanol, anhydrous, denatured, HPLC Grade, 90%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

100 mL, ChemSeal™ Bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
64-17-5
IUPAC Name
ethanol
Molecular Formula
C2H6O
InChI Key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES
CCO
Residue on Evaporation
5ppm
Residual water
0.1%

Description

Ethanol, anhydrous, denatured is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use