Tin foil, 0.1mm (0.004in) thick, 98.8% (metals basis)
Tin foil, 0.1mm (0.004in) thick, 98.8% (metals basis)
Tin foil, 0.1mm (0.004in) thick, 98.8% (metals basis)
Thermo Scientific Chemicals

Tin foil, 0.1mm (0.004in) thick, 98.8% (metals basis)

CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
產品號碼Quantity
044154.RG
亦稱為 044154-RG
30 x 150 cm
產品號碼 044154.RG
亦稱為 044154-RG
價格 (TWD)
-
Quantity:
30 x 150 cm
Request bulk or custom format
化學識別
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
檢視更多
規格Specification Sheet規格表
Appearance (Color)Silver
Assay from Supplier's CofA≥98.8% (metals basis)
FormFoil, 0.1mm (0.004in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only