Erbium powder, -40 mesh, 99.9% (REO)
Erbium powder, -40 mesh, 99.9% (REO)
Erbium powder, -40 mesh, 99.9% (REO)
Thermo Scientific Chemicals

Erbium powder, -40 mesh, 99.9% (REO)

CAS: 7440-52-0 | Er | 167.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 044169.06
also known as 044169-06
Price (EUR)
255,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
255,00
Each
Chemical Identifiers
CAS7440-52-0
IUPAC Nameerbium
Molecular FormulaEr
InChI KeyUYAHIZSMUZPPFV-UHFFFAOYSA-N
SMILES[Er]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only