Praseodymium powder, -40 mesh, 99.5% (REO)
Praseodymium powder, -40 mesh, 99.5% (REO)
Praseodymium powder, -40 mesh, 99.5% (REO)
Praseodymium powder, -40 mesh, 99.5% (REO)
Thermo Scientific Chemicals

Praseodymium powder, -40 mesh, 99.5% (REO)

CAS: 7440-10-0 | Pr | 140.91 g/mol
Have Questions?
Catalog NumberQuantityPackaging
044183.06
also known as 044183-06
5 gAmpule
Catalog number 044183.06
also known as 044183-06
Price (EUR)
219,00
Each
Add to cart
Quantity:
5 g
Packaging:
Ampule
Request bulk or custom format
Price (EUR)
219,00
Each
Add to cart
Chemical Identifiers
CAS7440-10-0
IUPAC Namepraseodymium
Molecular FormulaPr
InChI KeyPUDIUYLPXJFUGB-UHFFFAOYSA-N
SMILES[Pr]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.5% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only