What type of container is Rubidium 99.75% (metal basis) sold in?
Cat. No. 044214.xx Rubidium 99.75% (metal basis) is ampouled under Argon.
Cat. No. 010315.xx Rubidium 99.75% (metal basis) is vacuum sealed in a breakseal ampoule.
Cat. No. 010315.xx Rubidium 99.75% (metal basis) is vacuum sealed in a breakseal ampoule.