Thermo Scientific Chemicals

Tungsten slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.97% (metals basis), Thermo Scientific Chemicals

100 g
Thermo Scientific Chemicals

Tungsten slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.97% (metals basis), Thermo Scientific Chemicals

100 g
Quantity
Catalog number: 044252.22
also known as 044252-22
Price (USD)
Price: 163.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
7440-33-7
IUPAC Name
tungsten
Molecular Formula
W
InChI Key
WFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES
[W]
Assay from Supplier's CofA
≥99.97% (metals basis)
Appearance (Color)
Gray
Form
Slug, 6.35mm (0.25in) diameter.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only