Thermo Scientific Chemicals

Aqueous dispersant for multi-walled carbon nanotubes

Catalog number: 044276.AB
10 mL, Each
Thermo Scientific Chemicals

Aqueous dispersant for multi-walled carbon nanotubes

Catalog number: 044276.AB
10 mL, Each
Quantity
Catalog number: 044276.AB
also known as 044276-AB
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
Pale to dark yellow
Comment
Product is a dispersant package made for the production of stable aqueous suspensions of multi-walled carbon nanotubes. Nanotubes not included in product.
Comment
Density is approx. 1.05 g/mL
Form
Liquid

Description

For dispersing multi-walled carbon nanotubes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text