Manganese(II) perchlorate hexahydrate, 99.995% (metals basis)
Manganese(II) perchlorate hexahydrate, 99.995% (metals basis)
Manganese(II) perchlorate hexahydrate, 99.995% (metals basis)
Thermo Scientific Chemicals

Manganese(II) perchlorate hexahydrate, 99.995% (metals basis)

CAS: 15364-94-0 | Cl2H12MnO14 | 361.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 044318.09
also known as 044318-09
Price (TWD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS15364-94-0
IUPAC Namemanganese(2+) hexahydrate diperchlorate
Molecular FormulaCl2H12MnO14
InChI KeyAIFGVOMWPFMOCN-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Mn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
Manganese(II) perchlorate hexahydrate, is a good oxidizing agent and also acts as an intermediates in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only