Gallium pellets, 6mm dia, 99.99999% (metals basis)
Gallium pellets, 6mm dia, 99.99999% (metals basis)
Gallium pellets, 6mm dia, 99.99999% (metals basis)
Gallium pellets, 6mm dia, 99.99999% (metals basis)
Thermo Scientific Chemicals

Gallium pellets, 6mm dia, 99.99999% (metals basis)

CAS: 7440-55-3 | Ga | 69.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 044325.18
also known as 044325-18
Price (EUR)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS7440-55-3
IUPAC Namegallium
Molecular FormulaGa
InChI KeyGYHNNYVSQQEPJS-UHFFFAOYSA-N
SMILES[Ga]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.00001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only