Aluminum rod, 22mm (0.87in) dia, Puratronic™, 99.9999% metals basis)
Aluminum rod, 22mm (0.87in) dia, Puratronic™, 99.9999% metals basis)
Aluminum rod, 22mm (0.87in) dia, Puratronic™, 99.9999% metals basis)
Thermo Scientific Chemicals

Aluminum rod, 22mm (0.87in) dia, Puratronic™, 99.9999% metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Catalog NumberQuantity
044336.BQ
also known as 044336-BQ
5 cm
Catalog number 044336.BQ
also known as 044336-BQ
Price (USD)
513.65
Special offer
Online exclusive
Ends: 30-Jun-2026
604.00
Save 90.35 (15%)
Each
Quantity:
5 cm
Request bulk or custom format
Price (USD)
513.65
Special offer
Online exclusive
Ends: 30-Jun-2026
604.00
Save 90.35 (15%)
Each
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod
Appearance (Color)Silver
Total Metal Impurities0.0001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only