Trityl chloride, polymer-supported, 1% cross-linked, 100-200 mesh, 0.5-2.5 mmol/g on poly(styrene-divinylbenzene)
Trityl chloride, polymer-supported, 1% cross-linked, 100-200 mesh, 0.5-2.5 mmol/g on poly(styrene-divinylbenzene)
Trityl chloride, polymer-supported, 1% cross-linked, 100-200 mesh, 0.5-2.5 mmol/g on poly(styrene-divinylbenzene)
Trityl chloride, polymer-supported, 1% cross-linked, 100-200 mesh, 0.5-2.5 mmol/g on poly(styrene-divinylbenzene)
Thermo Scientific Chemicals

Trityl chloride, polymer-supported, 1% cross-linked, 100-200 mesh, 0.5-2.5 mmol/g on poly(styrene-divinylbenzene)

CAS: 76-83-5 | C19H15Cl | 278.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 044397.14
also known as 044397-14
Price (EUR)
868,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
868,00
Each
Add to cart
Specifications
CAS76-83-5
Chemical Name or MaterialTrityl chloride
ColorWhite
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Health Hazard 1H314-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only