Cyclohexyl methyl methylphosphonate, 99%, Thermo Scientific Chemicals
Cyclohexyl methyl methylphosphonate, 99%, Thermo Scientific Chemicals
Cyclohexyl methyl methylphosphonate, 99%, Thermo Scientific Chemicals
Cyclohexyl methyl methylphosphonate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclohexyl methyl methylphosphonate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044426.04
also known as 044426-04
Price (TWD)
-
Request A Quote
Quantity:
2 g
Request bulk or custom format
Specifications
CAS7040-52-0
Chemical Name or MaterialCyclohexyl methyl methylphosphonate
Formula Weight192.09
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Cyclohexyl methyl methylphosphonate or phosphonic acid is used as a micro-nano electronic materials have extended self-assembled monolayers (SAMs) beyond conventional gold/thiol systems. To expand the choice of the substrate used for the preparation of SAMs, the chemical functionalities in film-forming molecules can be altered by introducing phosphate or phosphonate groups. Such polar acidic molecules are capable of interacting with diverse metal-oxide surfaces (e.g., Al2O3, Ta2O5, NbO5, ZrO2 and TiO2) and form films with a similar degree of ordering as for alkyl thiol SAMs on gold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only