Scandium(III) 2,4-pentanedionate hydrate
Scandium(III) 2,4-pentanedionate hydrate
Scandium(III) 2,4-pentanedionate hydrate
Scandium(III) 2,4-pentanedionate hydrate
Thermo Scientific Chemicals

Scandium(III) 2,4-pentanedionate hydrate

CAS: 699012-88-9 | C15H21O6Sc | 342.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 044448.06
also known as 044448-06
Price (EUR)
415,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
415,00
Each
Chemical Identifiers
CAS699012-88-9
IUPAC Namescandium(3+) tris(2,4-dioxopentan-3-ide)
Molecular FormulaC15H21O6Sc
InChI KeyNYPXMGRKLWIPFL-UHFFFAOYSA-N
SMILES[Sc+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown to yellow
FormPowder
Water ContentReport Value
Assay from Suppliers CofASc(acac)3: Report Value
Assay from Suppliers CofAScandium REO: Report Value
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only