Specifications
CAS100-53-8
Chemical Name or MaterialBenzyl mercaptan
ColorPink to White
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more