Thermo Scientific Chemicals

Sodium hexafluoroaluminate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 044478.36
500 g, Each
Thermo Scientific Chemicals

Sodium hexafluoroaluminate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 044478.36
500 g, Each
Quantity
Catalog number: 044478.36
also known as 044478-36
Price (USD)
Price: 508.00
Online price: 431.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13775-53-6
IUPAC Name
aluminium(3+) trisodium hexafluoride
Molecular Formula
AlF6Na3
InChI Key
REHXRBDMVPYGJX-UHFFFAOYSA-H
SMILES
[F-].[F-].[F-].[F-].[F-].[F-].[Na+].[Na+].[Na+].[Al+3]
Total Metal Impurities
0.1% max.

Description

Sodium aluminum hexafluoride is used as a solvent (or flux) for electrolysis aluminum oxides such as bauxite, whitener for enamels and an opacifier for glass and in the industrial production of aluminum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text