What are the dimensions of the Polystyrene Test Film?
The test film is 19 mm in diameter and 38 microns thick. The holder is 5.08 cm (2 inches) x 10.2 cm (4 inches).
Search
View additional product information for Polystyrene Test Film - FAQs (044502.KT)
2 product FAQs found