Aluminum 2,4-pentanedionate, 99.995+% (metals basis)
Aluminum 2,4-pentanedionate, 99.995+% (metals basis)
Aluminum 2,4-pentanedionate, 99.995+% (metals basis)
Thermo Scientific Chemicals

Aluminum 2,4-pentanedionate, 99.995+% (metals basis)

CAS: 13963-57-0 | C15H21AlO6 | 324.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 044538.14
also known as 044538-14
Price (USD)
221.65
Online Exclusive
246.00
Save 24.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
221.65
Online Exclusive
246.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS13963-57-0
IUPAC Namealuminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC15H21AlO6
InChI KeyKILURZWTCGSYRE-LNTINUHCSA-K
SMILES[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.005%
Appearance (Color)White to pale yellow-green
Assay from Suppliers CofAReport metals analysis
FormPowder or crystals or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only