Aluminum etchant - FAQs
Aluminum etchant - FAQs
View additional product information for Aluminum etchant - FAQs (044581.LV, 044581.R0)
11 product FAQs found
What is the solubility of Aluminum etchant?
Aluminum etchant is miscible with water.
What is the flash point of Aluminum etchant?
Aluminum etchant is non-inflammable.
What is the specific gravity (25 degrees C) of Aluminum etchant?
The specific gravity (25 degrees C) of Aluminum etchant is 1.45.
What is the freezing point of Aluminum etchant?
The freezing point of Aluminum etchant is <0 degrees C.
What is the boiling point of Aluminum etchant?
The boiling point of Aluminum etchant is >100 degrees C.
What is the pH of Aluminum etchant?
The pH of Aluminum etchant is 1.0.
How do I reduce undercutting of an etchant?
Undercutting of the etchant can be reduced by increasing the rate of stirring or agitation.
Do I need to dilute an etchant?
No, it is a ready-to-use solution. Dilution will only be necessary if you want to reduce the etch rate.
How do I reduce the etch rate of an etchant?
The etch rate of the etchant can be reduced by adding 1 part deionized water to 2 parts etchant. This will reduce the etch rate approximately 50%.
How do I increase the etch rate of an etchant?
The etch rate of the etchant will be increased by increasing the rate of stirring or agitation.
What is the etch rate of Aluminum etchant?
Aluminum etchant has the following etch rate:
  • 25 degrees C, 30 Å/sec
  • 40 degrees C, 80 Å/sec
Note: Etch rate may vary slightly based on purity of aluminum.