Chromium etchant - FAQs
Chromium etchant - FAQs
View additional product information for Chromium etchant - FAQs (044582.LV, 044582.R0)
5 product FAQs found
How do I reduce undercutting of Chromium etchant?
Undercutting of Chromium etchant can be reduced by increasing the rate of stirring or agitation.
Do I need to dilute Chromium etchant before use?
No, Chromium etchant is a ready-to-use solution. Dilution will only be necessary if you want to reduce the etch rate.
How do I reduce the etch rate of Chromium etchant?
The etch rate of Chromium etchant can be reduced by adding 1 part deionized water to 2 parts etchant. This will reduce the etch rate approximately 50%.
How do I increase the etch rate of Chromium etchant?
The etch rate of Chromium etchant will approximately double with every 10 degrees Celsius increase in temperature. The etch rate can also be increased by increasing the rate of stirring or agitation.
What is the etch rate of Chromium etchant at 40 degrees Celsius?
The etch rate of Chromium etchant at 40 degrees Celsius is 40 Angstroms/sec.