Thermo Scientific Chemicals

Praseodymium(III, IV) oxide, 99% (REO), Thermo Scientific Chemicals

Catalog number: 044594.30
250 g, Each
Thermo Scientific Chemicals

Praseodymium(III, IV) oxide, 99% (REO), Thermo Scientific Chemicals

Catalog number: 044594.30
250 g, Each
Quantity
Catalog number: 044594.30
also known as 044594-30
Price (USD)
Price: 139.00
Online price: 120.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12037-29-5
IUPAC Name
hexapraseodymium(3+) undecaoxidandiide
Molecular Formula
O11Pr6
InChI Key
HPZIIFVSYNLWNX-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Total Rare Earth Oxide Impurities
1% max.

Description

Used in ceramic structures. Have a wide range of applications from producing clay bowls to advanced electronics. Praseodymium oxide is also useful in light weight structural components in aerospace and electrochemical applications such as fuel cells due to its ionic conductivity. Component of didymium glass which is a colorant for welder's goggles, also as important additive of Praseodymium yellow pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text