Thermo Scientific Chemicals

Tetrahydrofuran, anhydrous, 99.8+%, stab. with 0.025% BHT, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

100 mL, Each
Thermo Scientific Chemicals

Tetrahydrofuran, anhydrous, 99.8+%, stab. with 0.025% BHT, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

100 mL, Each
Quantity
Catalog number: 044608.AE
also known as 044608-AE
Price (USD)
Price: 48.70
Your Price:
Quantity
-

Chemical Identifiers

CAS
109-99-9
IUPAC Name
oxolane
Molecular Formula
C4H8O
InChI Key
WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES
C1CCOC1
Appearance (Color)
Colorless
Form
Liquid
Assay (GC)
≥99.8% (Corrected for water)
Comment
Inert gas used can be either Argon or Nitrogen
Water Content (Karl Fischer Titration)
≤50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only