2-Amino-5-bromo-4-methylpyridine, 97%
2-Amino-5-bromo-4-methylpyridine, 97%
2-Amino-5-bromo-4-methylpyridine, 97%
Thermo Scientific Chemicals

2-Amino-5-bromo-4-methylpyridine, 97%

CAS: 98198-48-2 | C6H7BrN2 | 187.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 044623.06
also known as 044623-06
Price (EUR)
98,40
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
98,40
Each
Chemical Identifiers
CAS98198-48-2
IUPAC Name2-amino-5-bromo-4-methylpyridin-1-ium
Molecular FormulaC6H8BrN2
InChI KeyJDNCMHOKWINDKI-UHFFFAOYSA-O
SMILESCC1=CC(N)=[NH+]C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Very pale cream to light brown
Assay (unspecified)> 96.0%
2-Amino-5-bromo-4-methylpyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only