Search
Buscar
View additional product information for Poly(acrylic acid), 25 wt% soln. in water - FAQs (044669.36, 044669.22)
1 product FAQs found
The average molecular weight of the Poly(acrylic acid), 25 wt% soln. in water is 240,000 g/mole.