Palladium Nickel gauze, 50 mesh woven from 0.1578mm (0.0062in) dia wire
Thermo Scientific Chemicals

Palladium Nickel gauze, 50 mesh woven from 0.1578mm (0.0062in) dia wire

Filtration
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
Catalog number 044698.FL
also known as 044698-FL
Price (EUR)
524,00
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (EUR)
524,00
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangePd:Ni; 95:5 wt%
Chemical Name or MaterialPalladium Nickel gauze
Health Hazard 1H317-H350-H373
Health Hazard 2GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
Palladium Nickel gauze is used in filtrations. It is also used as a capture gauze for platinum recovery in ammonia oxidation apparatus with a pressure of 4 atm (405 kPa) and a temperature of 8500C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only