Silicon oxide, catalyst support, high surface area
Silicon oxide, catalyst support, high surface area
Silicon oxide, catalyst support, high surface area
Thermo Scientific Chemicals

Silicon oxide, catalyst support, high surface area

Form: S.A. 250m2/g | CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 044740.36
also known as 044740-36
Price (USD)
149.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
149.00
Each
Specifications
AppearanceWhite pellets
CAS7631-86-9
ChEBICHEBI:30563
Chemical Name or MaterialSilicon oxide
ColorWhite
View more
Typical properties: S.A.250m^2/gSilicon oxide is used as a catalyst support material having a high surface area. It acts as a medium to which catalyst is affixed. Several metals such as platinum, rhodium, ruthenium, silver, copper, nickel, iron, manganese, cobalt, tungsten and gold are supported over silicon dioxide and used as a catalyst in synthetic chemistry for various applications. For example, silica-supported gold catalysts is used for carbon monoxide oxidation by a solution-based technique.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

Is Silicon oxide, catalyst support, high surface area sold in powder or pellet form?

Silicon oxide, catalyst support, high surface area comes in pellet form.