Acrilato de (perfluorociclohexil)metilo, 96 %, estabilizado con 100 ppm 4-metoxifenol, Thermo Scientific Chemicals
Acrilato de (perfluorociclohexil)metilo, 96 %, estabilizado con 100 ppm 4-metoxifenol, Thermo Scientific Chemicals
Acrilato de (perfluorociclohexil)metilo, 96 %, estabilizado con 100 ppm 4-metoxifenol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acrilato de (perfluorociclohexil)metilo, 96 %, estabilizado con 100 ppm 4-metoxifenol, Thermo Scientific Chemicals

CAS: 40677-94-9 | C10H5F11O2 | 366.13 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
Número de catálogo 044793.06
también denominado 044793-06
Precio (EUR)
90,20
Each
Cantidad:
5 g
Pedido a granel o personalizado
Precio (EUR)
90,20
Each
Identificadores químicos
CAS40677-94-9
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Colorless
Assay (GC)≥ 95.0%
FormLiquid
Commentstabilized with 50-100ppm 4-methoxyphenol
(Perfluorocyclohexyl)methyl acrylate is used in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only