Bis(trimethylsilyl)sulfide, tech.
Bis(trimethylsilyl)sulfide, tech.
Bis(trimethylsilyl)sulfide, tech.
Thermo Scientific Chemicals

Bis(trimethylsilyl)sulfide, tech.

CAS: 3385-94-2 | C6H18SSi2 | 178.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 044818.06
also known as 044818-06
Price (USD)
196.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
196.00
Each
Specifications
Beilstein1740046
CAS3385-94-2
Chemical Name or MaterialBis (trimethylsilyl)sulfide
ColorColorless
DOT InformationHazard Class: 3; Packaging Group: III
View more
Bis(trimethylsilyl)sulfide is used to convert oxides and chlorides into the corresponding sulfides. It has been used in the conversion of aldehydes and ketones to the corresponding thiones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only