Barium 2-ethylhexanoate in 2-ethylhexanoic acid, Ba ≈17.5%, Thermo Scientific Chemicals
Barium 2-ethylhexanoate in 2-ethylhexanoic acid, Ba ≈17.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium 2-ethylhexanoate in 2-ethylhexanoic acid, Ba ≈17.5%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number 044847.14
also known as 044847-14
Price (USD)/ Each
67.90
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
67.90
Add to cart
Chemical Identifiers
IUPAC Namebarium(2+) bis(2-ethylhexanoate)
Molecular FormulaC16H30BaO4
InChI KeyVJFFDDQGMMQGTQ-UHFFFAOYNA-L
SMILES[Ba++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Molecular Weight (g/mol)423.74
View more
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid
Appearance (Color)Clear to slightly hazy pale yellow to brown
Complexometric TitrationBa content 17.0-18.0%
Appearance (Color)Clear to slightly hazy pale yellow to brown
FormViscous liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only