Specifications
Chemical Name or MaterialInstrument Calibration (ICAL) Standard Solution B
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more