Iodine lump, ultra dry, 99.999% (metals basis)
Iodine lump, ultra dry, 99.999% (metals basis)
Iodine lump, ultra dry, 99.999% (metals basis)
Iodine lump, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals

Iodine lump, ultra dry, 99.999% (metals basis)

CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044857.14
also known as 044857-14
Price (USD)
130.65
Special offer
Online exclusive
154.00
Save 23.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
130.65
Special offer
Online exclusive
154.00
Save 23.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7553-56-2
IUPAC Namediiodine
Molecular FormulaI2
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILESII
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Iodine lump is used in OLED materials, pharmaceuticals, and the electronics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.