Gallium(III) ethoxide
Gallium(III) ethoxide
Gallium(III) ethoxide
Thermo Scientific Chemicals

Gallium(III) ethoxide

CAS: 2572-25-0 | C6H18GaO3 | 207.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 044859.06
also known as 044859-06
Price (USD)
352.65
Online Exclusive
392.00
Save 39.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
352.65
Online Exclusive
392.00
Save 39.35 (10%)
Each
Chemical Identifiers
CAS2572-25-0
IUPAC Nametris(ethanol) gallium
Molecular FormulaC6H18GaO3
InChI KeyJWISYUBLQXUTOS-UHFFFAOYSA-N
SMILES[Ga].CCO.CCO.CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.1%
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream or yellow
Gallium(III) ethoxide cohydrolysis with Te alkoxides on PMMA yields heat-mode erasable optical memory. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only