Ruthenium powder, -200 mesh, Premion™, 99.99+% (metals basis excluding Ca), Ca 100ppm max
Ruthenium powder, -200 mesh, Premion™, 99.99+% (metals basis excluding Ca), Ca 100ppm max
Ruthenium powder, -200 mesh, Premion™, 99.99+% (metals basis excluding Ca), Ca 100ppm max
Ruthenium powder, -200 mesh, Premion™, 99.99+% (metals basis excluding Ca), Ca 100ppm max
Thermo Scientific Chemicals

Ruthenium powder, -200 mesh, Premion™, 99.99+% (metals basis excluding Ca), Ca 100ppm max

CAS: 7440-18-8 | Ru | 101.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number 044879.14
also known as 044879-14
Price (USD)/ Each
2,226.65
Special offer
Online exclusive
2,619.00
Save 392.35 (15%)
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
2,226.65
Special offer
Online exclusive
2,619.00
Save 392.35 (15%)
Add to cart
Chemical Identifiers
IUPAC Nameruthenium
Molecular FormulaRu
InChI KeyKJTLSVCANCCWHF-UHFFFAOYSA-N
SMILES[Ru]
Molecular Weight (g/mol)101.07
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)Ca ≤ 100ppm
Total Metal Impurities≤0.01%
Particle Size-200 mesh
FormPowder
Assay (special)Ru 99.8-100.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only